COMH4211A0C5 Combi-Blocks, Inc.

Benzyl 2-(isopropylcarbamoyl)pyrrolidine-1-carboxylate

  • Cas Number : 1219372-00-5
  • Molecular Weight : 290.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2171

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2171
Availability details for
Quantity
  • Availability
  • Add to Cart