COMH4211A0C6 Combi-Blocks, Inc.

benzyl 2-(cyclopentylcarbamoyl)pyrrolidine-1-carboxylate

  • Molecular Weight : 316.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2172

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2172
Availability details for
Quantity
  • Availability
  • Add to Cart