COMH4211A0CA Combi-Blocks, Inc.

1-Bromo-2-chloro-4-methoxy-3-nitrobenzene

  • MDL number MFCD30834310
  • Cas Number : 2092776-07-1
  • Molecular Weight : 266.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2409

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2409
Availability details for
Quantity
  • Availability
  • Add to Cart