COMH4211A0CD Combi-Blocks, Inc.

3-Phenyl-2H-pyrazole-4-carbonitrile

  • MDL number MFCD26637057
  • Cas Number : 5898-81-7
  • Molecular Weight : 169.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2459

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2459
Availability details for
Quantity
  • Availability
  • Add to Cart