COMH4211A0CE Combi-Blocks, Inc.

2-Bromo-N-isopropylpropanamide

  • MDL number MFCD15206799
  • Cas Number : 54537-46-1
  • Molecular Weight : 194.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2461

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2461
Availability details for
Quantity
  • Availability
  • Add to Cart