COMH4211A0D0 Combi-Blocks, Inc.

2-Bromo-1-(pyrrolidin-1-yl)propan-1-one

  • MDL number MFCD13173089
  • Cas Number : 54537-48-3
  • Molecular Weight : 206.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2463

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2463
Availability details for
Quantity
  • Availability
  • Add to Cart