COMH4211A0D1 Combi-Blocks, Inc.

2-Bromo-1-(piperidin-1-yl)propan-1-one

  • MDL number MFCD15207121
  • Cas Number : 2620-14-6
  • Molecular Weight : 220.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2464

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2464
Availability details for
Quantity
  • Availability
  • Add to Cart