COMH4211A0D2 Combi-Blocks, Inc.

2-Bromo-1-(morpholin-4-yl)propan-1-one

  • MDL number MFCD10697873
  • Cas Number : 2620-13-5
  • Molecular Weight : 222.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2465

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2465
Availability details for
Quantity
  • Availability
  • Add to Cart