COMH4211A0D3 Combi-Blocks, Inc.

2-Bromo-1-(4-methylpiperazin-1-yl)propan-1-one

  • MDL number MFCD13874871
  • Cas Number : 70060-39-8
  • Molecular Weight : 235.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2466

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2466
Availability details for
Quantity
  • Availability
  • Add to Cart