COMH4211A0D4 Combi-Blocks, Inc.

tert-Butyl 4-(2-bromopropanoyl)piperazine-1-carboxylate

  • MDL number MFCD32065696
  • Cas Number : 873089-06-6
  • Molecular Weight : 321.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2467

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2467
Availability details for
Quantity
  • Availability
  • Add to Cart