COMH4211A0D6 Combi-Blocks, Inc.

2-Bromo-N-(2-methoxyethyl)propanamide

  • MDL number MFCD15206701
  • Cas Number : 321861-49-8
  • Molecular Weight : 210.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2469

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2469
Availability details for
Quantity
  • Availability
  • Add to Cart