COMH4211A0D7 Combi-Blocks, Inc.

3,4-Dibromo-1H-indazole

  • MDL number MFCD07781594
  • Cas Number : 885521-68-6
  • Molecular Weight : 275.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2470

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2470
Availability details for
Quantity
  • Availability
  • Add to Cart