COMH4211A0D8 Combi-Blocks, Inc.

tert-Butyl 4-(2-methylphenyl)piperazine-1-carboxylate

  • MDL number MFCD11872561
  • Molecular Weight : 276.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2473

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2473
Availability details for
Quantity
  • Availability
  • Add to Cart