COMH4211A0DF Combi-Blocks, Inc.

tert-Butyl N-tert-butylcarbamate

  • MDL number MFCD21105433
  • Cas Number : 71872-03-2
  • Molecular Weight : 173.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2496

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2496
Availability details for
Quantity
  • Availability
  • Add to Cart