COMH4211A0E0 Combi-Blocks, Inc.

tert-butyl N-(2-methoxyethyl)carbamate

  • MDL number MFCD21105446
  • Cas Number : 30751-70-3
  • Molecular Weight : 175.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2497

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2497
Availability details for
Quantity
  • Availability
  • Add to Cart