COMH4211A0E2 Combi-Blocks, Inc.

5-Fluorochromen-4-one

  • MDL number MFCD24713280
  • Cas Number : 1386253-18-4
  • Molecular Weight : 164.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2520

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2520
Availability details for
Quantity
  • Availability
  • Add to Cart