COMH4211A0E4 Combi-Blocks, Inc.

2,2-Dimethylpropyl 4-fluorobenzenesulfonate

  • MDL number MFCD32065702
  • Cas Number : 81390-32-1
  • Molecular Weight : 246.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2522

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2522
Availability details for
Quantity
  • Availability
  • Add to Cart