COMH4211A0E5 Combi-Blocks, Inc.

(2-Isopropylphenyl)methanol

  • MDL number MFCD09999704
  • Cas Number : 21190-34-1
  • Molecular Weight : 150.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2524

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2524
Availability details for
Quantity
  • Availability
  • Add to Cart