COMH4211A0E8 Combi-Blocks, Inc.

2-Chloro-n-cyclopropylquinazolin-4-amine

  • MDL number MFCD03306415
  • Cas Number : 524033-55-4
  • Molecular Weight : 219.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8929

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8929
Availability details for
Quantity
  • Availability
  • Add to Cart