COMH4211A0E9 Combi-Blocks, Inc.

6-Bromo-N-methyl-3-nitropyridin-2-amine

  • MDL number MFCD31672605
  • Cas Number : 924293-34-5
  • Molecular Weight : 232.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2532

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2532
Availability details for
Quantity
  • Availability
  • Add to Cart