COMH4211A0F0 Combi-Blocks, Inc.

4-(2,4-Dichlorophenoxy)piperidine

  • MDL number MFCD08687543
  • Cas Number : 367501-06-2
  • Molecular Weight : 246.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2542

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2542
Availability details for
Quantity
  • Availability
  • Add to Cart