COMH4211A0F1 Combi-Blocks, Inc.

5-Bromo-2-(piperidin-3-yloxy)pyridine

  • MDL number MFCD11877787
  • Cas Number : 1250803-96-3
  • Molecular Weight : 257.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2543

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?LD-2543
Availability details for
Quantity
  • Availability
  • Add to Cart