COMH4211A0FD Combi-Blocks, Inc.

4-Chloro-o-xylene

  • MDL number MFCD00000596
  • Cas Number : 615-60-1
  • Molecular Weight : 140.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-0178

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-0178
Availability details for
Quantity
  • Availability
  • Add to Cart