COMH4211A166 Combi-Blocks, Inc.

(S)-(-)-1-(P-Tolyl)ethylamine

  • MDL number MFCD00145246
  • Cas Number : 27298-98-2
  • Molecular Weight : 135.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-3867

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-3867
Availability details for
Quantity
  • Availability
  • Add to Cart