COMH4211A174 Combi-Blocks, Inc.

S-3-Hydroxytetrahydrothiophene

  • MDL number MFCD00798268
  • Cas Number : 79107-75-8
  • Molecular Weight : 104.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4586

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4586
Availability details for
Quantity
  • Availability
  • Add to Cart