COMH4211A17B Combi-Blocks, Inc.

(S)-2-Aminooctane

  • MDL number MFCD01091021
  • Cas Number : 34566-04-6
  • Molecular Weight : 129.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4758

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-4758
Availability details for
Quantity
  • Availability
  • Add to Cart