COMH4211A184 Combi-Blocks, Inc.

4-Chloro-3-(trifluoromethyl)pyridine

  • MDL number MFCD02183559
  • Cas Number : 387824-63-7
  • Molecular Weight : 181.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-5299

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-5299
Availability details for
Quantity
  • Availability
  • Add to Cart