COMH4211A188 Combi-Blocks, Inc.

3-Hydroxyindole

  • MDL number MFCD02683932
  • Cas Number : 480-93-3
  • Molecular Weight : 133.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-5426

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-5426
Availability details for
Quantity
  • Availability
  • Add to Cart