COMH4211A1D4 Combi-Blocks, Inc.

1-Benzyl-3-methyl pyrrolidine-3-carboxylic acid

  • MDL number MFCD05662803
  • Cas Number : 181114-74-9
  • Molecular Weight : 219.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-benzyl-3-methyl-3-pyrrolidinecarboxylic acid
Inchi Code 1S/C13H17NO2/c1-13(12(15)16)7-8-14(10-13)9-11-5-3-2-4-6-11/h2-6H,7-10H2,1H3,(H,15,16)
Inchi Key MSUXGUJKSWGNTC-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-6476

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-6476
Availability details for
Quantity
  • Availability
  • Add to Cart