COMH4211A1DE Combi-Blocks, Inc.

3-(Benzyloxy)-o-toluidine

  • MDL number MFCD06858675
  • Cas Number : 65361-82-2
  • Molecular Weight : 213.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-7060

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-7060
Availability details for
Quantity
  • Availability
  • Add to Cart