COMH4211A1E7 Combi-Blocks, Inc.

4-Amino-piperidine-4-carboxylic acid, HCl

  • MDL number MFCD08271757
  • Molecular Weight : 180.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-7657

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-7657
Availability details for
Quantity
  • Availability
  • Add to Cart