COMH4211A1F0 Combi-Blocks, Inc.

3-Butyl-5-methyl isoxazol-4-carboxylic acid ethyl ester

  • MDL number MFCD13185143
  • Cas Number : 1239465-20-3
  • Molecular Weight : 211.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 3-butyl-5-methyl-4-isoxazolecarboxylate
Inchi Code 1S/C11H17NO3/c1-4-6-7-9-10(8(3)15-12-9)11(13)14-5-2/h4-7H2,1-3H3
Inchi Key UPGNSGNBQCREJB-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8440

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-8440
Availability details for
Quantity
  • Availability
  • Add to Cart