COMH4211A1F5 Combi-Blocks, Inc.

1-(3,4-Difluorophenyl)cyclohexanecarbonitrile

  • MDL number MFCD11036792
  • Molecular Weight : 221.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9752

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9752
Availability details for
Quantity
  • Availability
  • Add to Cart