COMH4211A1F8 Combi-Blocks, Inc.

1-(Spiro[indoline-3,4'-piperidin]-1-yl)ethanone hcl

  • MDL number MFCD18711456
  • Cas Number : 167484-19-7
  • Molecular Weight : 266.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9912

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9912
Availability details for
Quantity
  • Availability
  • Add to Cart