COMH4211A203 Combi-Blocks, Inc.

2-Bromo-5-chloro-3-methoxypyridine

  • MDL number MFCD09835246
  • Cas Number : 127561-71-1
  • Molecular Weight : 222.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OS-2317

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OS-2317
Availability details for
Quantity
  • Availability
  • Add to Cart