COMH4211A204 Combi-Blocks, Inc.

2-Bromo-3-methoxypyridine-n-oxide

  • MDL number MFCD09839263
  • Cas Number : 104819-48-9
  • Molecular Weight : 204

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OS-2326

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OS-2326
Availability details for
Quantity
  • Availability
  • Add to Cart