COMH4211A205 Combi-Blocks, Inc.

1-Bromo-3-(2,2,2-trifluoro-ethylsulfanyl)-benzene

  • MDL number MFCD06657737
  • Cas Number : 850349-30-3
  • Molecular Weight : 271.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OS-2409

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OS-2409
Availability details for
Quantity
  • Availability
  • Add to Cart