COMH4211A206 Combi-Blocks, Inc.

2-Amino-5-bromo-3-methylthiopyrazine

  • MDL number MFCD08460058
  • Cas Number : 87444-38-0
  • Molecular Weight : 220.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OS-2437

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OS-2437
Availability details for
Quantity
  • Availability
  • Add to Cart