COMH4211A207 Combi-Blocks, Inc.

9-Bromo-1,1,7,7-tetramethyl-2,3,6,7-tetrahydro-1h,5h-pyrido[3,2,1-ij]quinoline

  • MDL number MFCD09835257
  • Cas Number : 1259033-32-3
  • Molecular Weight : 308.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OS-2457

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OS-2457
Availability details for
Quantity
  • Availability
  • Add to Cart