COMH4211A211 Combi-Blocks, Inc.

1-Bromo-3-methoxy-5-propoxybenzene

  • MDL number MFCD32065708
  • Cas Number : 2365419-96-9
  • Molecular Weight : 245.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-1421

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-1421
Availability details for
Quantity
  • Availability
  • Add to Cart