COMH4211A212 Combi-Blocks, Inc.

1-bromo-3-[(4-methoxyphenyl)methoxy]benzene

  • MDL number MFCD11127626
  • Cas Number : 944469-62-9
  • Molecular Weight : 293.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-1703

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-1703
Availability details for
Quantity
  • Availability
  • Add to Cart