COMH4211A216 Combi-Blocks, Inc.

tert-Butyl N-isopropylcarbamate

  • MDL number MFCD02380471
  • Cas Number : 51170-55-9
  • Molecular Weight : 159.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2079

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2079
Availability details for
Quantity
  • Availability
  • Add to Cart