COMH4211A219 Combi-Blocks, Inc.

6-bromo-4,4-dimethyl-2,3-dihydro-1-benzothiopyran

  • MDL number MFCD15144704
  • Cas Number : 112110-44-8
  • Molecular Weight : 257.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2518

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2518
Availability details for
Quantity
  • Availability
  • Add to Cart