COMH4211A24B Combi-Blocks, Inc.

1-[(tert-Butoxy)carbonyl]-3-(trifluoromethyl)piperidine-4-carboxylic acid

  • MDL number MFCD11975952
  • Cas Number : 1260649-60-2
  • Molecular Weight : 297.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4366

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4366
Availability details for
Quantity
  • Availability
  • Add to Cart