COMH4211A24D Combi-Blocks, Inc.

1-{[(3-Bromophenyl)methane]sulfonyl}-4-methylpiperazine

  • MDL number MFCD28396553
  • Cas Number : 1160924-46-8
  • Molecular Weight : 333.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4694

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4694
Availability details for
Quantity
  • Availability
  • Add to Cart