COMH4211A24F Combi-Blocks, Inc.

8-(trifluoromethoxy)quinolin-5-ol

  • MDL number MFCD18417173
  • Cas Number : 1261749-13-6
  • Molecular Weight : 229.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4721

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4721
Availability details for
Quantity
  • Availability
  • Add to Cart