COMH4211A250 Combi-Blocks, Inc.

8-Ethoxyquinolin-5-ol

  • MDL number MFCD24719970
  • Molecular Weight : 189.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4722

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4722
Availability details for
Quantity
  • Availability
  • Add to Cart