COMH4211A253 Combi-Blocks, Inc.

4-Chloro-1-isocyano-2-(trifluoromethyl)benzene

  • MDL number MFCD02664549
  • Cas Number : 601465-68-3
  • Molecular Weight : 205.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4727

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4727
Availability details for
Quantity
  • Availability
  • Add to Cart