COMH4211A254 Combi-Blocks, Inc.

4-Bromo-2-fluoro-1-isocyanobenzene

  • MDL number MFCD04117591
  • Cas Number : 730964-93-9
  • Molecular Weight : 200.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4728

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4728
Availability details for
Quantity
  • Availability
  • Add to Cart