COMH4211A256 Combi-Blocks, Inc.

tert-Butyl (9aR)-4-oxo-hexahydropiperazino[2,1-c]morpholine-8-carboxylate

  • MDL number MFCD28003010
  • Cas Number : 1382034-27-6
  • Molecular Weight : 256.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4730

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4730
Availability details for
Quantity
  • Availability
  • Add to Cart